Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, w/o Adapters, Pump or Controller

Evaporates up to 6 samples on one compact unit

Specifications

Certifications CE, CSA, ISO I4000
Capacity (Metric) 10 to 160mL
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
For Use With (Application) High throughput labs requiring multiple sample processing
Model P-6
View More Specs

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Products
Catalog Number Mfr. No. Includes Price Quantity    

05-404-717

 
buchi™
MP22199S10
Safety shield, 1L recieving flask CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

05-404-720

 
buchi™
MP22199S20
Safety shield, 2L recieving flask CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Certifications CE, CSA, ISO I4000
Capacity (Metric) 10 to 160mL
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
For Use With (Application) High throughput labs requiring multiple sample processing
Model P-6
Temperature Range (Metric) 95°C
Electrical Requirements 100-120V 50/60Hz
Height (English) Footprint 15.7 in.
Height (Metric) Footprint 40cm
Hertz 50/60Hz
Length (English) Footprint 10.2 in.
Length (Metric) Footprint 26cm
Maximum Speed Range 0 to 485rpm
Temperature Range (English) 203°F
Voltage 100/120V
Wattage 800w
Weight (English) 46.3 lb.
Weight (Metric) 21kg

Samples are heated separately in cylindrical transparent water baths. Boiling retardation is prevented by a vigorous vortex induced by a thorough horizontal orbital movement of the heating plate. A vacuum manifold with individual sample connection conducts the vapor to the condenser assembly of the rotary evaporator.

Efficient

  • Ideal for high throughput labs requiring multiple sample processing
  • Use the same condenser and vacuum assembly for two instruments, minimizing sample handling
  • Samples can be prepared in advance and stored in the evaporation rack
  • Footprint measures only 26 × 40cm
Safe
  • Each sample is independently connected to the vacuum cover to eliminate cross-contamination
  • Reduces environmental impact as compared to nitrogen blow-down evaporators
  • Samples are visible throughout evaporation
  • Meets ISO I4000 guidelines
  • For 100-120V 50/60Hz
Versatile
  • Accommodates a wide variety of vessel types with up to 30mL working volume
  • Processes a high number of extraction samples quickly using modern pressurized extraction techniques such as PSE or ASE
  • Evaporation of parallel reaction samples is achieved in the same reaction tube
  • Optional PE frits allow adsorption on silica for subsequent chromatographic purification
  • Screw capped test tubes containing chromatographic samples can be evaporated simultaneously with minimal user supervision

Other configurations are available. Contact your Customer Service Representative for details.