Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, V-700/V-855, Sec. Condenser, w/o Adapters


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP22199S22

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-722

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Height (Metric) 40cm
Hertz 50/60Hz
Voltage 100/120V
Weight (English) 46.3 lb.
Weight (Metric) 21kg
Width (Metric) 27cm
Width (English) 10.6 in.
Temperature Range (Metric) 95°C
For Use With (Application) High throughput labs requiring multiple sample processing
Maximum Speed Range 0 to 485rpm
Temperature Range (English) 203°F
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Height (English) 5.74 in.
Model P-6
Wattage 800w
Volume (English) 67.5 oz.
Electrical Requirements 100-120V 50/60Hz
Height (Metric) Footprint 40cm
Length (English) Footprint 10.2 in.
Volume (Metric) 2000mL
Length (English) 5.74 in.
Length (Metric) 40cm
Capacity (Metric) 10 to 160mL
Height (English) Footprint 15.7 in.
Includes Safety shield, V-700 pump, V-855 controller, woulff bottle, secondary condenser, 2L receiving flask
Length (Metric) Footprint 26cm