BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, V-700/V-855, Sec. Condenser, w/o Adapter
Evaporates up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP21199S22
 View more versions of this product
Catalog No. 05-404-774
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
Multivapor P-6 Parallel Evaporation System With S Condenser
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
21kg
220/240V
40cm
5.74 in.
High throughput labs requiring multiple sample processing
40cm
203°F
10 to 160mL
15.7 in.
27cm
40cm
10.6 in.
CE, CSA, ISO I4000
0 to 485rpm
10.2 in.
95°C
5.74 in.
800w
26cm
220-240V 50/60Hz
67.5 oz.
2000mL
Safety shield, V-700 pump, V-855 controller, woulff bottle, secondary condenser, 2L receiving flask
50/60Hz
46.3 lb.
P-6