Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, V-700/V-855, Sec. Condenser for 220-240V


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21099S22

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-748

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Voltage 220/240V
Maximum Speed Range 0 to 485rpm
Hertz 50/60Hz
Width (English) 10.6 in.
Height (English) 5.74 in.
Weight (English) 46.3 lb.
Length (Metric) Footprint 26cm
Model P-6
Electrical Requirements 220-240V 50/60Hz
Volume (English) 67.5 oz.
Length (Metric) 40cm
Height (Metric) Footprint 40cm
Temperature Range (English) 203°F
Height (English) Footprint 15.7 in.
For Use With (Application) High throughput labs requiring multiple sample processing
Length (English) Footprint 10.2 in.
Includes V-700 pump, V-855 controller, woulff bottle, 2L receiving flask
Volume (Metric) 2000mL
Wattage 800w
Capacity (Metric) 10 to 160mL
Weight (Metric) 21kg
Width (Metric) 27cm
Length (English) 5.74 in.
Height (Metric) 40cm
Temperature Range (Metric) 95°C