Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, Safety Shield, w/o Pump or Controller


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP22101S20

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-707

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Maximum Speed Range 0 to 485rpm
Height (Metric) 40cm
Length (English) 5.74 in.
Length (Metric) 40cm
Voltage 100/120V
Weight (English) 46.3 lb.
Weight (Metric) 21kg
Width (English) 10.6 in.
Model P-6
Temperature Range (English) 203°F
Volume (English) 67.5 oz.
Volume (Metric) 2000mL
Temperature Range (Metric) 95°C
For Use With (Application) High throughput labs requiring multiple sample processing
Holds 150mL Tube
Includes Safety shield, 2L recieving flask
Length (English) Footprint 10.2 in.
Length (Metric) Footprint 26cm
Wattage 800w
Electrical Requirements 100-120V 50/60Hz
For Use With (Equipment) 150mL tubes
Hertz 50/60Hz
Width (Metric) 27cm
Capacity (Metric) 10 to 160mL
Height (English) Footprint 15.7 in.
Height (Metric) Footprint 40cm
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Height (English) 5.74 in.