Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, Safety shield, Tube, Adapters


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP22101S11

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-705

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Weight (Metric) 21kg
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Maximum Speed Range 0 to 485rpm
Volume (English) 34 oz.
Height (Metric) 40cm
Height (English) 5.74 in.
Weight (English) 46.3 lb.
Length (Metric) 40cm
Height (Metric) Footprint 40cm
Wattage 800w
For Use With (Equipment) 150mL tubes
Electrical Requirements 100-120V 50/60Hz
Holds 150mL Tube
Height (English) Footprint 15.7 in.
Includes Safety shield, V-700 pump, V-801 controller, woulff bottle, 1L recieving flask
Temperature Range (English) 203°F
Length (English) Footprint 10.2 in.
Volume (Metric) 1000mL
Capacity (Metric) 10 to 160mL
For Use With (Application) High throughput labs requiring multiple sample processing
Length (Metric) Footprint 26cm
Length (English) 5.74 in.
Width (Metric) 27cm
Width (English) 10.6 in.
Voltage 100/120V
Hertz 50/60Hz
Temperature Range (Metric) 95°C
Model P-6