BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser,Adapter. w/o Pump, controller for 220-240V
Evaporates up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP21101S10
 View more versions of this product
Catalog No. 05-404-756
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
Multivapor P-6 Parallel Evaporation System With S Condenser
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
34 oz.
5.74 in.
50/60Hz
10.2 in.
40cm
21kg
P-6
150mL tubes
High throughput labs requiring multiple sample processing
40cm
10 to 160mL
Safety shield, 1L recieving flask
5.74 in.
CE, CSA, ISO I4000
0 to 485rpm
27cm
40cm
220/240V
10.6 in.
95°C
26cm
220-240V 50/60Hz
150mL Tube
15.7 in.
800w
203°F
1000mL
46.3 lb.