Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser,Adapter. w/o Pump, controller for 220-240V


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21101S20

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-759

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor P-6 Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Holds 150mL Tube
Maximum Speed Range 0 to 485rpm
Height (Metric) 40cm
Length (Metric) 40cm
Length (English) 5.74 in.
Width (English) 10.6 in.
Temperature Range (Metric) 95°C
Weight (Metric) 21kg
Height (English) 5.74 in.
Model P-6
For Use With (Equipment) 150mL tubes
Electrical Requirements 220-240V 50/60Hz
Volume (English) 67.5 oz.
Height (English) Footprint 15.7 in.
Wattage 800w
Volume (Metric) 2000mL
Capacity (Metric) 10 to 160mL
Length (English) Footprint 10.2 in.
Height (Metric) Footprint 40cm
Temperature Range (English) 203°F
Includes Safety shield, 2L recieving flask
For Use With (Application) High throughput labs requiring multiple sample processing
Length (Metric) Footprint 26cm
Weight (English) 46.3 lb.
Voltage 220/240V
Hertz 50/60Hz
Width (Metric) 27cm