Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/o Pump, Controller and Glassware/Adapter


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21099C20

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-752

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporators w/o Pump, Controller and Glassware/Adapter
Certifications CE, CSA, ISO I4000
Maximum Speed Range 0 to 485rpm
Weight (English) 46.3 lb.
Width (English) 10.6 in.
Length (Metric) 40cm
Hertz 50/60Hz
Width (Metric) 27cm
Height (Metric) 40cm
Weight (Metric) 21kg
Length (Metric) Footprint 26cm
Includes 2L receiving flask
Electrical Requirements 220-240V 50/60Hz
Volume (English) 67.5 oz.
Temperature Range (English) 203°F
Length (English) Footprint 10.2 in.
Wattage 800w
Volume (Metric) 2000mL
Height (Metric) Footprint 40cm
Height (English) Footprint 15.7 in.
Capacity (Metric) 10 to 160mL
For Use With (Application) High throughput labs requiring multiple sample processing
Length (English) 5.74 in.
Height (English) 5.74 in.
Temperature Range (Metric) 95°C
Model P-6
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Voltage 220/240V