Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/o Pump and Controller for 220-240V


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21001C10

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-736

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporators w/o Pump and Controller
Certifications CE, CSA, ISO I4000
Holds 150mL Tube
Volume (English) 0.26 gal.
Length (Metric) 40cm
Voltage 220/240V
Width (English) 10.6 in.
Model P-6
Temperature Range (Metric) 95°C
Capacity (Metric) 10 to 160mL
For Use With (Application) High throughput labs requiring multiple sample processing
Includes Buchi tube, tube adapters, 1L recieving flask
Height (Metric) Footprint 40cm
Maximum Speed Range 0 to 485rpm
Length (English) Footprint 10.2 in.
Length (Metric) Footprint 26cm
Temperature Range (English) 203°F
Wattage 800w
Volume (Metric) 1000mL
Electrical Requirements 220-240V 50/60Hz
Height (Metric) 40cm
Hertz 50/60Hz
Length (English) 5.74 in.
Weight (English) 46.3 lb.
Weight (Metric) 21kg
Width (Metric) 27cm
Height (English) Footprint 15.7 in.
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Height (English) 5.74 in.