BUCHI™ Multivapor™ P-6 Parallel Evaporators w/o Pump and Controller for 220-240V
Evaporates up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP21001C20
 View more versions of this product
Catalog No. 05-404-739
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
CE, CSA, ISO I4000
5.74 in.
50/60Hz
40cm
46.3 lb.
27cm
P-6
10 to 160mL
15.7 in.
Buchi tube, tube adapters, 2L recieving flask
0 to 485rpm
26cm
800w
2000mL
220-240V 50/60Hz
Multivapor™ Parallel Evaporators w/o Pump and Controller
40cm
5.74 in.
220/240V
21kg
10.6 in.
95°C
High throughput labs requiring multiple sample processing
150mL Tube
40cm
10.2 in.
203°F
0.53 gal.
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)