Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators with Safety Shield and w/o Pump, Controller


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21101C10

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-762

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Parallel Evaporators with Safety Shield and w/o Pump, Controller
Certifications CE, CSA, ISO I4000
Maximum Speed Range 0 to 485rpm
Volume (English) 34 oz.
Holds 150mL Tube
Height (English) 5.74 in.
Height (Metric) 40cm
Hertz 50/60Hz
Voltage 220/240V
Weight (Metric) 21kg
Length (English) 5.74 in.
Temperature Range (Metric) 95°C
Height (English) Footprint 15.7 in.
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 220-240V 50/60Hz
Includes Safety shield, 1L receiving flask
Temperature Range (English) 203°F
Height (Metric) Footprint 40cm
For Use With (Application) High throughput labs requiring multiple sample processing
Capacity (Metric) 10 to 160mL
Volume (Metric) 1000mL
Length (English) Footprint 10.2 in.
Wattage 800w
Length (Metric) Footprint 26cm
Weight (English) 46.3 lb.
Width (Metric) 27cm
Width (English) 10.6 in.
Model P-6
Length (Metric) 40cm