Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators with S and Secondary Condenser


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP22001S22

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-680

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Parallel Evaporators with S and Secondary Condenser
Certifications CE, CSA, ISO I4000
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Holds 150mL Tube
Volume (English) 67.5 oz.
Height (Metric) 40cm
Width (Metric) 27cm
Weight (Metric) 21kg
Width (English) 10.6 in.
Temperature Range (Metric) 95°C
Height (English) 5.74 in.
Capacity (Metric) 10 to 160mL
Length (English) Footprint 10.2 in.
For Use With (Equipment) 150mL tubes
Electrical Requirements 120V 50/60Hz
Maximum Speed Range 0 to 485rpm
Temperature Range (English) 203°F
Length (Metric) Footprint 26cm
Wattage 800w
Height (Metric) Footprint 40cm
Includes V-700 pump, V-855 controller, woulff bottle, secondary condenser, 2L Receiving Flask
Height (English) Footprint 15.7 in.
For Use With (Application) High throughput labs requiring multiple sample processing
Volume (Metric) 2000mL
Length (English) 5.74 in.
Voltage 100/120V
Hertz 50/60Hz
Model P-6
Length (Metric) 40cm
Weight (English) 46.3 lb.