BUCHI™ Multivapor™ P-6 Parallel Evaporators with S Condenser, w/o Safety Shield, Adapters
Evaporates up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP22099S22
 View more versions of this product
Catalog No. 05-404-695
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
Multivapor™ Parallel Evaporation System With S Condenser
100/120V
V-700 pump, V-855 controller, woulff bottle, secondary condenser, 2L recieveing flask
40cm
5.74 in.
15.7 in.
120V 50/60Hz
67.5 oz.
2000mL
High throughput labs requiring multiple sample processing
26cm
50/60Hz
10.6 in.
46.3 lb.
CE, CSA, ISO I4000
10.2 in.
27cm
5.74 in.
95°C
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
0 to 485rpm
40cm
800w
203°F
10 to 160mL
21kg
40cm
P-6