BUCHI™ Multivapor™ P-6 Parallel Evaporators with S Condenser and w/o Pump or Controller
Evaporates up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP22001S20
 View more versions of this product
Catalog No. 05-404-678
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
Parallel Evaporators with S Condenser and w/o Pump or Controller
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
67.5 oz.
40cm
5.74 in.
40cm
2000mL
150mL tubes
40cm
10 to 160mL
26cm
15.7 in.
800w
46.3 lb.
P-6
CE, CSA, ISO I4000
0 to 485rpm
50/60Hz
100/120V
21kg
27cm
95°C
100-120V 50/60Hz
203°F
2L Receiving Flask
10.2 in.
High throughput labs requiring multiple sample processing
5.74 in.
10.6 in.