Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators with S Condenser w/o Adapters, Pump or Controller


Evaporate up to 6 samples on one compact unit

Manufacturer: buchi™  MP22099S20

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-693

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Parallel Evaporators with S Condenser w/o Adapters, Pump or Controller
Certifications CE, CSA, ISO I4000
Height (Metric) 40cm
Hertz 50/60Hz
Weight (English) 46.3 lb.
Width (Metric) 27cm
Width (English) 10.6 in.
Model P-6
Temperature Range (Metric) 95°C
Capacity (Metric) 10 to 160mL
For Use With (Application) High throughput labs requiring multiple sample processing
Height (English) Footprint 15.7 in.
Includes 2L Receiving Flask
Height (Metric) Footprint 40cm
Maximum Speed Range 0 to 485rpm
Length (Metric) Footprint 26cm
Temperature Range (English) 203°F
Volume (English) 67.5 oz.
Volume (Metric) 2000mL
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 100-120V 50/60Hz
Height (English) 5.74 in.
For Use With (Equipment) 2L receiving flask
Length (English) 5.74 in.
Length (Metric) 40cm
Voltage 100/120V
Weight (Metric) 21kg
Length (English) Footprint 10.2 in.
Wattage 800w