Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators with S Condenser, V-700 Pump, V-855 Controller


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP22101S12

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-706

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Maximum Speed Range 0 to 485rpm
Holds 150mL Tube
Volume (English) 34 oz.
Hertz 50/60Hz
Height (English) 5.74 in.
Voltage 100/120V
Width (English) 10.6 in.
For Use With (Application) High throughput labs requiring multiple sample processing
For Use With (Equipment) 150mL tubes
Electrical Requirements 100-120V 50/60Hz
Includes Safety shield, V-700 pump, V-855 controller, woulff bottle, secondary condenser, 1L recieving flask
Capacity (Metric) 10 to 160mL
Height (English) Footprint 15.7 in.
Wattage 800w
Volume (Metric) 1000mL
Length (English) Footprint 10.2 in.
Temperature Range (English) 203°F
Height (Metric) Footprint 40cm
Length (Metric) Footprint 26cm
Length (English) 5.74 in.
Weight (Metric) 21kg
Height (Metric) 40cm
Weight (English) 46.3 lb.
Model P-6
Length (Metric) 40cm
Width (Metric) 27cm
Temperature Range (Metric) 95°C