Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporator w/S Condenser, Adapters, w/o Pump, Controller for 220-240V


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21001S10

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-730

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Holds 150mL Tube
Maximum Speed Range 0 to 485rpm
Volume (English) 34 oz.
Hertz 50/60Hz
Length (Metric) 40cm
Voltage 220/240V
Weight (English) 46.3 lb.
Weight (Metric) 21kg
Width (Metric) 27cm
Capacity (Metric) 10 to 160mL
For Use With (Application) High throughput labs requiring multiple sample processing
Length (English) Footprint 10.2 in.
Temperature Range (English) 203°F
Wattage 800w
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 220-240V 50/60Hz
Height (English) 5.74 in.
For Use With (Equipment) 150mL tubes
Height (English) Footprint 15.7 in.
Includes 1L Receiving Flask
Volume (Metric) 1000mL
Height (Metric) 40cm
Length (English) 5.74 in.
Width (English) 10.6 in.
Height (Metric) Footprint 40cm
Length (Metric) Footprint 26cm
Model P-6
Temperature Range (Metric) 95°C