Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporator w/S Condenser, Adapters, V-700/V-855 for 220-240V


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21001S12

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-732

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Length (English) 5.74 in.
Length (Metric) 40cm
Width (English) 10.6 in.
Holds 150mL Tube
Maximum Speed Range 0 to 485rpm
Height (English) 5.74 in.
Capacity (Metric) 10 to 160mL
Height (English) Footprint 15.7 in.
Includes V-700 pump, V-855 controller, woulff bottle, secondary condenser, 1L recieving flask
Height (Metric) Footprint 40cm
Temperature Range (English) 203°F
Volume (English) 34 oz.
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 220-240V 50/60Hz
For Use With (Equipment) 150mL tubes
For Use With (Application) High throughput labs requiring multiple sample processing
Length (English) Footprint 10.2 in.
Length (Metric) Footprint 26cm
Volume (Metric) 1000mL
Height (Metric) 40cm
Hertz 50/60Hz
Voltage 220/240V
Weight (English) 46.3 lb.
Weight (Metric) 21kg
Width (Metric) 27cm
Temperature Range (Metric) 95°C
Wattage 800w
Model P-6