BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with Rotavapor Interface for 220-240V
Evaporate up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP21099R00
 View more versions of this product
Catalog No. 05-404-755
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
CE, CSA, ISO I4000
5.74 in.
50/60Hz
40cm
46.3 lb.
27cm
P-6
10 to 160mL
15.7 in.
40cm
10.2 in.
203°F
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Multivapor™ Parallel Evaporation System With Rotavapor Interface
40cm
5.74 in.
220/240V
21kg
10.6 in.
95°C
Sample processing in high-throughput labs
Not Applicable
0 to 485rpm
26cm
800w
220-240V 50/60Hz