BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with Rotavapor Interface for 220-240V
Evaporate up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP21199R00
 View more versions of this product
Catalog No. 05-404-781
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
Multivapor™ Parallel Evaporation System With Rotavapor Interface
21kg
5.74 in.
Safety shield
High throughput labs requiring multiple sample processing
220-240V 50/60Hz
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
15.7 in.
26cm
203°F
27cm
5.74 in.
40cm
CE, CSA, ISO I4000
0 to 485rpm
50/60Hz
220/240V
800w
95°C
10 to 160mL
40cm
10.2 in.
40cm
10.6 in.
P-6
46.3 lb.