Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with Rotavapor Interface for 100-120V


Evaporate up to 6 samples on one compact unit

Manufacturer: buchi™  MP22199R00

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-729

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With Rotavapor Interface
Certifications CE, CSA, ISO I4000
Maximum Speed Range 0 to 485rpm
Voltage 100/120V
Weight (Metric) 21kg
Height (English) 5.74 in.
Length (English) 5.74 in.
Weight (English) 46.3 lb.
Height (Metric) Footprint 40cm
Temperature Range (Metric) 95°C
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 110-120V 50/60Hz
Length (English) Footprint 10.2 in.
Height (English) Footprint 15.7 in.
Wattage 800w
For Use With (Application) High throughput labs requiring multiple sample processing
Includes Safety shield
Capacity (Metric) 10 to 160mL
Temperature Range (English) 203°F
Length (Metric) Footprint 26cm
Length (Metric) 40cm
Height (Metric) 40cm
Hertz 50/60Hz
Model P-6
Width (English) 10.6 in.
Width (Metric) 27cm