Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with Rotavapor Interface for 100-120V


Evaporate up to 6 samples on one compact unit

Manufacturer: buchi™  MP22101R00

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-716

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With Rotavapor Interface
Certifications CE, CSA, ISO I4000
Hertz 50/60Hz
Length (English) 5.74 in.
Width (Metric) 27cm
Width (English) 10.6 in.
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Voltage 100/120V
Weight (English) 46.3 lb.
Weight (Metric) 21kg
Capacity (Metric) 10 to 160mL
Height (English) Footprint 15.7 in.
Holds 150mL Tube
Includes Safety shield, adapter
Height (Metric) Footprint 40cm
Maximum Speed Range 0 to 485rpm
Length (Metric) Footprint 26cm
Temperature Range (English) 203°F
Wattage 800w
Electrical Requirements 120V 50/60Hz
For Use With (Equipment) 150mL tubes
Height (Metric) 40cm
Length (Metric) 40cm
Model P-6
Temperature Range (Metric) 95°C
For Use With (Application) High throughput labs requiring multiple sample processing
Length (English) Footprint 10.2 in.
Height (English) 5.74 in.