Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with C Condenser for 100-120V


Evaporate up to 6 samples on one compact unit

Manufacturer: buchi™  MP22001C20

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-685

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With C Condenser
Certifications CE, CSA, ISO I4000
Weight (English) 46.3 lb.
Width (Metric) 27cm
Holds 150mL Tube
Maximum Speed Range 0 to 485rpm
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Height (Metric) 40cm
Voltage 100/120V
Width (English) 10.6 in.
Length (English) Footprint 10.2 in.
Length (English) 5.74 in.
Weight (Metric) 21kg
Model P-6
Temperature Range (Metric) 95°C
Capacity (Metric) 10 to 160mL
Length (Metric) Footprint 26cm
Electrical Requirements 100-120V 50/60Hz
For Use With (Equipment) 150mL tubes, 2L receiving flask
For Use With (Application) High throughput labs requiring multiple sample processing
Height (English) Footprint 15.7 in.
Height (Metric) Footprint 40cm
Temperature Range (English) 203°F
Wattage 800w
Volume (Metric) 2000mL
Hertz 50/60Hz
Length (Metric) 40cm
Height (English) 5.74 in.