Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with C Condenser for 100-120V


Evaporate up to 6 samples on one compact unit

Manufacturer: buchi™  MP22001C12

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-684

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With C Condenser
Certifications CE, CSA, ISO I4000
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Length (Metric) 40cm
Voltage 100/120V
Height (English) 5.74 in.
Weight (English) 46.3 lb.
Width (Metric) 27cm
Temperature Range (Metric) 95°C
Model P-6
For Use With (Equipment) 150mL tubes, 1L receiving flask
Electrical Requirements 100-120V 50/60Hz
Maximum Speed Range 0 to 485rpm
Holds 150mL Tube
Includes V-700 pump, V-855 controller, woulff bottle, secondary condenser
Temperature Range (English) 203°F
Capacity (Metric) 10 to 160mL
Length (English) Footprint 10.2 in.
Length (Metric) Footprint 26cm
Volume (Metric) 1000mL
Height (Metric) Footprint 40cm
Height (English) Footprint 15.7 in.
For Use With (Application) High throughput labs requiring multiple sample processing
Wattage 800w
Width (English) 10.6 in.
Hertz 50/60Hz
Weight (Metric) 21kg
Length (English) 5.74 in.
Height (Metric) 40cm