Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with C Condenser for 100-120V


Evaporate up to 6 samples on one compact unit

Manufacturer: buchi™  MP22199C22

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-728

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With C Condenser
Certifications CE, CSA, ISO I4000
Height (Metric) 40cm
Hertz 50/60Hz
Length (Metric) 40cm
Weight (English) 46.3 lb.
Width (English) 10.6 in.
Model P-6
Temperature Range (Metric) 95°C
Height (English) Footprint 15.7 in.
Maximum Speed Range 0 to 485rpm
Electrical Requirements 100-120V 50/60Hz
Height (English) 5.74 in.
For Use With (Equipment) 2L receiving flask
Capacity (Metric) 10 to 160mL
Includes Safety shield, V-700 pump, V-855 controller, woulff bottle, secondary condenser
Height (Metric) Footprint 40cm
Length (English) Footprint 10.2 in.
Length (Metric) Footprint 26cm
Temperature Range (English) 203°F
Wattage 800w
Volume (Metric) 2000mL
Length (English) 5.74 in.
Voltage 100/120V
Weight (Metric) 21kg
Width (Metric) 27cm
For Use With (Application) High throughput labs requiring multiple sample processing
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)