BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with C Condenser for 100-120V
Evaporate up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP22199C21
 View more versions of this product
Catalog No. 05-404-727
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
Multivapor™ Parallel Evaporation System With C Condenser
0 to 485rpm
5.74 in.
10 to 160mL
5.74 in.
100-120V 50/60Hz
10.2 in.
40cm
Safety shield, V-700 pump, V-801 controller, woulff bottle
800w
21kg
40cm
40cm
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
CE, CSA, ISO I4000
100/120V
46.3 lb.
10.6 in.
2L receiving flask
26cm
203°F
15.7 in.
2000mL
High throughput labs requiring multiple sample processing
27cm
50/60Hz
P-6
95°C