Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with C Condenser for 100-120V


Evaporate up to 6 samples on one compact unit

Manufacturer: buchi™  MP22101C12

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-712

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With C Condenser
Certifications CE, CSA, ISO I4000
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Holds 150mL Tube
Maximum Speed Range 0 to 485rpm
Length (Metric) 40cm
Width (Metric) 27cm
Includes Safety shield, V-700 pump, V-855 controller, woulff bottle, secondary condenser
Voltage 100/120V
Width (English) 10.6 in.
Length (English) 5.74 in.
Height (English) 5.74 in.
Length (English) Footprint 10.2 in.
Model P-6
Temperature Range (Metric) 95°C
For Use With (Equipment) 150mL tubes, 1L receiving flask
Electrical Requirements 100-120V 50/60Hz
Capacity (Metric) 10 to 160mL
Height (Metric) Footprint 40cm
Height (English) Footprint 15.7 in.
Temperature Range (English) 203°F
Length (Metric) Footprint 26cm
Wattage 800w
For Use With (Application) High throughput labs requiring multiple sample processing
Volume (Metric) 1000mL
Weight (Metric) 21kg
Weight (English) 46.3 lb.
Height (Metric) 40cm
Hertz 50/60Hz