Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with C Condenser for 100-120V


Evaporate up to 6 samples on one compact unit

Manufacturer: buchi™  MP22099C21

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-701

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporation System With C Condenser
Certifications CE, CSA, ISO I4000
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Width (Metric) 27cm
Weight (English) 46.3 lb.
Width (English) 10.6 in.
Voltage 100/120V
Length (Metric) 40cm
Temperature Range (Metric) 95°C
Height (Metric) 40cm
Capacity (Metric) 10 to 160mL
Model P-6
For Use With (Equipment) 2L receiving flask
Electrical Requirements 100-120V 50/60Hz
Maximum Speed Range 0 to 485rpm
Wattage 800w
Volume (Metric) 2000mL
For Use With (Application) High throughput labs requiring multiple sample processing
Temperature Range (English) 203°F
Includes V-700 pump, V-801 controller, woulff bottle
Height (Metric) Footprint 40cm
Length (English) Footprint 10.2 in.
Height (English) Footprint 15.7 in.
Length (Metric) Footprint 26cm
Height (English) 5.74 in.
Length (English) 5.74 in.
Hertz 50/60Hz
Weight (Metric) 21kg