BUCHI™ Large Capacity Evaporating Flasks
Standard glass flasks for use with various Rotavapor™ Evaporator models
$2,054.78 - $3,637.85
Specifications
Material Standard Glass
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Volume (Metric) For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Volume (Metric) For Use With (Equipment) Price Quantity  
05-402-410 BUCHI™
027468
20000mL 10L R-153 and R-220 rotary evaporator Each for $2,253.25
05-402-411 BUCHI™
027469
10000mL 6L R-153 and R-220 rotary evaporator Each for $2,054.78
05-402-412 BUCHI™
027470
6000mL 20L R-153 and R-220 rotary evaporator Each for $2,054.78
05-402-696 BUCHI™
001156
6000mL 20L R-151 and R-152 rotary evaporator Each for $2,284.02
05-402-715 BUCHI™
002608
10000mL 6L R-151 and R-152 rotary evaporator Each for $2,115.22
05-402-722 BUCHI™
003777
20000mL 20L R-185 and R-186 rotary evaporator Each for $3,637.85
05-402-771 BUCHI™
011899
20000mL 20L R-151 rotary evaporator Each for $3,121.89
05-404-259 BUCHI™
027010
20000mL 10L R-152 rotary evaporator Each for $2,668.42
Specifications
Description
  • Inert industrial glass (borosilicate 3.3, ISO/DIN 3585)
  • Tension relieved at 540°C
Description & Specifications
Specifications
Standard Glass