BUCHI™ Glass Columns C-690
Manufacturer:  BUCHI™ 044044
 View more versions of this product
Catalog No. 05-402-627
Specifications Description & Specifications
Specifications
Preparative separations together with the C-600 Sepacore™ line of chromatography separators
Borosilicate Glass
230mm
290psi
Glass column
49mm