BUCHI™ Glass Columns C-690
$2,248.75 - $9,175.80
Specifications
For Use With Preparative separations together with the C-600 Sepacore™ line of chromatography separators
Material Borosilicate Glass
Type Glass column
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Inner Max. Pressure Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Max. Pressure Length (Metric) Price Quantity  
05-402-617 BUCHI™
044031
15mm 725psi 230mm Each for $2,248.75
05-402-618 BUCHI™
044032
15mm 725psi 460mm Each for $2,302.05
05-402-619 BUCHI™
044033
15mm 725psi 920mm Each for $2,679.40
05-402-621 BUCHI™
044036
26mm 580psi 230mm Each for $2,302.05
05-402-622 BUCHI™
044037
26mm 580psi 460mm Each for $2,377.18
05-402-623 BUCHI™
044038
26mm 580psi 920mm Each for $2,728.76
05-402-625 BUCHI™
044041
36mm 435psi 460mm Each for $2,540.75
05-402-624 BUCHI™
044040
36mm 435psi 230mm Each for $2,465.53
05-402-626 BUCHI™
044042
36mm 435psi 920mm Each for $3,071.91
05-402-627 BUCHI™
044044
49mm 290psi 230mm Each for $2,874.49
05-402-628 BUCHI™
044045
49mm 290psi 460mm Each for $3,090.73
05-402-629 BUCHI™
044046
49mm 290psi 920mm Each for $3,408.02
05-402-630 BUCHI™
044048
70mm 218psi 230mm Each for $4,644.31
05-402-631 BUCHI™
044049
70mm 218psi 460mm Each for $4,759.47
05-402-632 BUCHI™
044050
70mm 218psi 920mm Each for $5,847.69
05-402-633 BUCHI™
044052
100mm 145psi 230mm Each for $8,376.67
05-402-634 BUCHI™
044053
100mm 145psi 460mm Each for $9,175.80
05-402-635 BUCHI™
044054
100mm 145psi 920mm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
Preparative separations together with the C-600 Sepacore™ line of chromatography separators
Borosilicate Glass
Glass column