Please login to your online account to display your discounted pricing
BUCHI™ Glass Column MPLC
Manufacturer: buchi™  019674
 View more versions of this product
Catalog No. 05-402-806
  • / Each
Description & Specifications
Specifications
Material Borosilicate Glass
Length (Metric) 230mm
Diameter (Metric) Inner 32mm
Type MPLC Glass column
Column Type Sepacore™ C-690 MPLC Glass column
Stationary Phase Sepacore™ C-690
Packing Material Sepacore™ C-690