Please login to your online account to display your discounted pricing
BUCHI™ Flanged Evaporating Flasks for Reflux Systems
Flanged flasks for use with Rotavapor™ evaporators
$685.47 - $1,051.18
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    
05-402-749
buchi™
011146
250mL Each for $685.47
05-402-750
buchi™
011147
500mL Each for $685.47
05-402-751
buchi™
011148
1000mL Each for $768.69
05-402-752
buchi™
011149
2000mL Each for $856.29
05-402-753
buchi™
011150
3000mL Each for $1,051.18
05-402-765
buchi™
011607
100mL Each for $762.11
Description & Specifications
  • Standard tapered joint: 24/40
  • Volume: 100–3000mL