BUCHI Kugelrohr and Drying Glass Ovens
Gentle fractional distillation or sample drying with full sample visibility
Supplier: BUCHI 046600
Description
- Electrically conductive glass tubing allows full visibility of samples at any time
- Gentle sample treatment provides high yield
- Digital display depicts operating temperature
- Multiple pre-settings provide ease of use
- High efficiency saves energy
- Compact design saves space
Specifications
100 to 250 mL | |
Drying Oven | |
30 x 41 x 30 cm (11.8 x 16.1 x 11.8 in.) (D x W x H) | |
50/60 Hz |
40°C to 300°C ±5°C | |
B-585 Drying | |
100/230 V |