BUCHI™ Drying Flasks for Rotary Evaporators
Special shaped flasks for powder drying
Manufacturer:  BUCHI™ 000453
 View more versions of this product
Catalog No. 05-402-669
Specifications Description & Specifications
Specifications
29.2/32
CE
Rotary evaporator
Standard Glass
60°C
1000mL
1000mL
Powder Drying Flask
Plastic and Glass
29.2/32