Please login to your online account to display your discounted pricing
BUCHI™ Distillation Spiders for Five-Flask Distillations
Process up to 5 samples in a single parallel evaporation process
$3,170.91 - $3,591.13
Products
Catalog Number Mfr. No. Price Quantity    
05-402-700
buchi™
001332
Each for $3,189.18
05-402-701
buchi™
001333
Each for $3,189.18
05-402-758
buchi™
011574
Each for $3,170.91
05-402-759
buchi™
011575
Each for $3,591.13
Description & Specifications
  • 5 positions
  • Holds 50 or 100mL cylindrical flasks
  • STJ 29/32 or 24/40