Please login to your online account to display your discounted pricing
BUCHI™ Distillation Spiders for Five-Flask Distillations
Process up to 5 samples in a single parallel evaporation process
Manufacturer: buchi™  001333
 View more versions of this product
Catalog No. 05-402-701
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) 5 flask distillation
Product Type Distillation Spider
Taper 29.2/32
Capacity (Metric) 100mL