Please login to your online account to display your discounted pricing
BUCHI™ Beaker-Style Drying Flasks for Rotary Evaporators
Manufacturer: buchi™  034767
 View more versions of this product
Catalog No. 05-404-334
  • / Each
Description & Specifications
Specifications
Temperature (Metric) Operating 60°C
Capacity (Metric) 150mL
For Use With (Equipment) Rotary evaporator
Product Type Drying Flask
Material Standard Glass
Material (Coating) Plastic and Glass
Shape Pear
Taper 29/32