Please login to your online account to display your discounted pricing

BU 239 hydrochloride, Tocris

Potent, highly selective I2 ligand

$172.26 - $726.06

Chemical Identifiers

Chemical Name or Material BU 239 hydrochloride
Assay Percent Range >98%
Molecular Formula C11H10N4·HCl
Formula Weight 234.69
Synonym 2-(4, 5-Dihydroimidazol-2-yl)quinoxaline hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

072650

 
r & d systems
RDS072650
50mg Each for $726.06

072610

 
r & d systems
RDS072610
10mg Each for $172.26
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 234.69
Chemical Name or Material BU 239 hydrochloride
Assay Percent Range >98%
Molecular Formula C11H10N4·HCl
CAS 1217041-98-9
Synonym 2-(4, 5-Dihydroimidazol-2-yl)quinoxaline hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature