Please login to your online account to display your discounted pricing

BU 226 hydrochloride, Tocris


Potent, highly selective I2 ligand

Quantity 50mg

Manufacturer: r & d systems  RDS109150

 View more versions of this product

Catalog No. 109150

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material BU 226 hydrochloride
Quantity 50mg
CAS 1186195-56-1
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C12H11N3·HCl
Formula Weight 233.7
Synonym 2-(4, 5-Dihydroimidazol-2-yl)isoquinoline hydrochloride

Specifications

Chemical Name or Material BU 226 hydrochloride
Assay Percent Range >98%
Molecular Formula C12H11N3·HCl
Formula Weight 233.7
Synonym 2-(4, 5-Dihydroimidazol-2-yl)isoquinoline hydrochloride
Accessories

Store at Room Temperature