Please login to your online account to display your discounted pricing

BU 224 hydrochloride, Tocris

Potent, selective I2 ligand; Putative antagonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material BU 224 hydrochloride
Assay Percent Range >98%
Molecular Formula C12H11N3·HCl
Formula Weight 233.7
Synonym 2-(4, 5-Dihydroimidazol-2-yl)quinoline hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

072550

 
r & d systems
RDS072550
50mg Each for $697.40

072510

 
r & d systems
RDS072510
10mg Each for $168.15
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 233.7
Assay Percent Range >98%
Molecular Formula C12H11N3·HCl
CAS 205437-64-5
Chemical Name or Material BU 224 hydrochloride
Synonym 2-(4, 5-Dihydroimidazol-2-yl)quinoline hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature