Please login to your online account to display your discounted pricing

BTX Harvard Microslides for Electrofusion Chambers


Manufacturer: btx genetronics  450105

 View more versions of this product

Catalog No. BTX453

  • / Each
 

Description & Specifications

Specifications

Field Type Homogeneous
Material Stainless Steel
Size (English) Gap 0.12 in.
Size (Metric) Gap 3.2mm
Volume (English) 0.012 oz.
Volume (Metric) 650μL
For Use With (Application) Embryo manipulation, nuclear transfer, cell fusion, hybridoma production, plant protoplast fusion
For Use With (Equipment) ECM 830, ECM 2001 generator