Please login to your online account to display your discounted pricing
BTX™ Harvard Electroporation Chambers - Cuvettes Plus™
Field strengths up to 25kV/cm are possible when these cuvets are used with BTX generators
$382.15 - $382.15
Specifications
Disposable Yes
For Use With (Equipment) BTX generators
Includes Sterile pipet for removing the sample after electroporation
Sterile Yes
View More Specs
Products
Catalog Number Mfr. No. Closure Color For Use With (Application) Volume (Metric) Width (Metric) Gap Price Quantity    
BTX610
btx genetronics
450124
Gray Bacteria 90μL 1mm Pack of 50 Each for $382.15
BTX620
btx genetronics
450125
Blue Bacteria, mammalian, plant 400μL 2mm Pack of 50 Each for $382.15
BTX640
btx genetronics
450126
Yellow Mammalian, plant 800μL 4mm Pack of 50 Each for $382.15
Description & Specifications
Specifications
Disposable Yes
For Use With (Equipment) BTX generators
Includes Sterile pipet for removing the sample after electroporation
Sterile Yes
  • With imbedded aluminum electrodes and round, color-coded caps
  • Gamma irradiated for sterility
  • Each package of 50 individually wrapped cuvets includes a sterile transfer pipet for removing the sample after electroporation