Please login to your online account to display your discounted pricing

BTX™-Harvard Apparatus HT 25-,96-Well Electroporation Plates


For use with ECM 630 and ECM 830 Electroporation Systems

Manufacturer: btx genetronics  450463

 View more versions of this product

Catalog No. BTX45-0463

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Size (English) Gap 0.15 in.
Volume (English) 0.008 oz.
Electroporation Chamber 25-well Electroporation Plate
Size (Metric) Gap 4mm
Volume (Metric) 250μL
For Use With (Equipment) Gemini X2 96-well electroporation system
Quantity 6/Pk.
For Use With (Application) Transfection of mammalian cell, transformation of bacteria and yeast, siRNA and cDNA library screening