Please login to your online account to display your discounted pricing
BTX™-Harvard Apparatus HT 25-,96-Well Electroporation Plates
For use with ECM 630 and ECM 830 Electroporation Systems
Manufacturer: btx genetronics  450462
 View more versions of this product
Catalog No. BTX45-0462
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) Gemini X2 96-well electroporation system
Volume (Metric) 250μL
Size (Metric) Gap 4mm
Size (English) Gap 0.15 in.
Electroporation Chamber 25-well Electroporation Plate
Quantity 1/Pk.
Volume (English) 0.008 oz.
For Use With (Application) siRNA transfection, library studies and stem cell project