Please login to your online account to display your discounted pricing

BTX™-Harvard Apparatus ECM™ 630 Exponential Decay Wave Electroporator

Broad capacitance and resistance range supports a wide variety of in vitro and in vivo applications

Specifications

Capacitance (HV Mode) 25, 50μF
Capacitance (LV Mode) 1, 25 to 3275μF
Certifications/Compliance CE
Charging Time 24 sec.
Depth (English) 12.25 in.
View More Specs
Includes:  The ECM 630 Electroporation system includes Exponential Decay Pulse Generator; 30 sterile, disposable cuvets (1, 2, and 4mm gap sizes); cuvet safety stand; 20-position numbered cuvet rack
Products
Catalog Number Mfr. No. For Use With (Application) Price Quantity    

BTX6300

 
btx genetronics
450001
Transformation of bacteria and yeast, transfection of mammalian cells, transfection of plant tissue and protoplast, electroporation of medaka oocytes and insect cells CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTX630

 
btx genetronics
450051
Allows the researcher full flexibility in choosing a wide range of time constants CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Capacitance (HV Mode) 25, 50μF
Capacitance (LV Mode) 1, 25 to 3275μF
Certifications/Compliance CE
Charging Time 24 sec.
Depth (English) 12.25 in.
Depth (Metric) 311mm
Display LCD
Hertz 50/60Hz
Voltage 100/240VAC
Electrical Requirements 100/240VAC 50/60Hz
Electroporation Chamber Safety stand
Height (English) 5.5 in.
Height (Metric) 140mm
Interface Digital user
Pulse Form Exponential decay
Pulse Length Range 625μsec. to 78msec.
Voltage Range (HV Mode) 50/2500VDC
Voltage Range (LV Mode) 10/500VDC
Weight (English) 10 lb.
Weight (Metric) 4.53kg
Width (English) 12.5 in.
Width (Metric) 318mm

Ideal for gram positive and gram negative bacteria, yeast, fungi, and other microorganism transformation in HV mode; optimized for a broad range of mammalian cell lines such as ES cells, fibroblasts, and epithelial cells in the LV mode. Novel applications include transdermal drug/gene delivery and intact plant gene delivery.

  • Delivers a maximum of 6000A in low-voltage mode; Precision Pulse™ System provides the user with excellent control of the time constant
  • Features BTX Power Platform technology, digital user interface, built-in monitoring of voltage and pulse length, automatic charge and pulse sequence, single rotary encoder, programming options, online menu, and protocol storage
  • Duplicates over 200 ECM 600 protocols; the additional ECM 630 resistor selection NONE allows researchers to reproduce protocols from competitive systems lacking resistor settings or reporting unlimited resistance

Applications:

Ideal for bacterial, yeast, fungal, and other microorganism transformation in HV mode; optimized for a broad range of mammalian cell lines such as ES cells, fibroblasts, and epithelial cells in the LV mode; novel applications include transdermal drug/gene delivery and intact plant gene delivery