missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BSTFA with 1% TMCS N,O-Bis(trimethylsilyl)trifluoroacetamide, ProteoSpec™, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIRMB1202T10X1C2

Catalog No. MB1202T1012



Description

Description

  • Modify amino acid side chains to block side chain interactions

Certifications

CofA

Specifications

Specifications

0.97
Colorless
25561-30-2
2924
Soluble in water
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
257.403
257.4g/mol
≥99%
40°C to 50°C
10 x 1g
C8H18F3NOSi2
acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl
XCOBLONWWXQEBS-GHXNOFRVSA-N
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
9601896
Liquid

Chemical Identifiers

25561-30-2
257.403
acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
C8H18F3NOSi2
XCOBLONWWXQEBS-GHXNOFRVSA-N
9601896
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
Safety and Handling

Safety and Handling

ShelfLife : 12 months

storageNote1 : Stable at room temperature

Recommended Storage : 15° to 30°C

Documents