Please login to your online account to display your discounted pricing

Thermo Scientific™ BseMI (BsrDI)


Cut at GCAATG(2/0)^ sites with BseMI (BsrDI) restriction enzyme, which performs best at 55°C in R buffer (Isoschizomers: Bse3DI, BsrDI).

Manufacturer: thermo scientific™  ER1262

 View more versions of this product

Catalog No. FERER1262

  • / Each
 

Description & Specifications

Specifications

Product Name BseMI (BsrDI)
Incubator Temperature 55°C
Quantity 500U
Concentration 5U/µL
Product Name BseMI (BsrDI)
Quantity 500U

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities