missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ BSA Silylation Reagent

Produce volatile, stable TMS derivatives quickly and quantitatively under mild conditions with Thermo Scientific™ BSA Silylation Reagent.

$911.03 - $911.03

Specifications

For Use With GC
Hazards Corrosive, Flammable
Standard Components N-(Trimethylsilyl)acetamide
Standard Concentration 3 to 5%
Storage 2 - 8°C
View More Specs

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI38836 Thermo Scientific™
TS38836
10 x 1mL Pack of 10 Each for $911.03
Specifications

Description

Derivatize analytes quickly and quantitatively under mild conditions using Thermo Scientific™ BSA Silylation Reagent. This reagent reacts readily with a wide range of compounds, including alcohols, amines, carboxylic acids, phenols, steroids, biogenic amines, and alkaloids to form volatile, stable TMS derivatives for GC.

Recommended Use

Not recommended for use with carbohydrates or very low molecular weight compounds

Description & Specifications

Specifications

GC
N-(Trimethylsilyl)acetamide
2 - 8°C
Corrosive, Flammable
3 to 5%